Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione, 97%

Manufacturer:  Alfa Aesar™ A1207706

Quantity 5g

 View more versions of this product

Catalog No. AAA1207706


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

17587-22-3
296.19
UN1224
7,7-Dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedione; Fod-H
C10H11F7O2
MFCD00000435
2056991
1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione

Specifications

53°C (127°F)
5g
C10H11F7O2
MFCD00000435
2056991
1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione
56° to 57°C (10mmHg)
17587-22-3
296.19
UN1224
7,7-Dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedione; Fod-H
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit