Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,2-Tetrafluoroethane, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
811-97-2,67-56-1
C2H2F4
102.032
LVGUZGTVOIAKKC-UHFFFAOYSA-N
13129
1,1,1,2-tetrafluoroethane
C(C(F)(F)F)F
811-97-2,67-56-1
C2H2F4
102.032
LVGUZGTVOIAKKC-UHFFFAOYSA-N
13129
1,1,1,2-tetrafluoroethane
C(C(F)(F)F)F

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,1,1,2-Tetrafluoroethane811-97-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
CAS Min % 0.1,99.9
CAS Max % 0.1,99.9
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Solvent or Matrix Methanol-P&T
Quantity 1mL
Chemical Name or Material 1,1,1,2-Tetrafluoroethane

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only