Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, +99.5%, Pure, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
2.5kg
Packaging:
Glass Bottle
920-66-1
C3H2F6O
168.038
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol,hexafluoroisopropanol,hfip,hexafluoro-2-propanol,hexafluoroisopropyl alcohol,bis trifluoromethyl methanol,1,1,1,3,3,3-hexafluoroisopropanol,2h-hexafluoroisopropanol,2-propanol, 1,1,1,3,3,3-hexafluoro,1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O
920-66-1
C3H2F6O
168.038
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol,hexafluoroisopropanol,hfip,hexafluoro-2-propanol,hexafluoroisopropyl alcohol,bis trifluoromethyl methanol,1,1,1,3,3,3-hexafluoroisopropanol,2h-hexafluoroisopropanol,2-propanol, 1,1,1,3,3,3-hexafluoro,1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O
Water 0.3% max.
Infrared Spectrum Authentic
Specific Gravity 1.596
Packaging Glass Bottle
Boiling Point 59.0°C
Refractive Index 1.2740 to 1.2770
Density 1.5960g/mL
Flash Point >100°C
Vapor Pressure 102mmHg at 20°C
Melting Point -4.0°C
Color Colorless
Quantity 25g
CAS Min % 99.5
CAS Max % 100.0
Assay Percent Range 99.5+%
Linear Formula (CF3)2CHOH
UN Number 3265
Solubility Information Solubility in water: 1000g/L (25°C).
Formula Weight 168.04
Physical Form Liquid
Percent Purity ≥99.5%
Grade Pure
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less

GHS P Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger