Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass bottle
920-66-1
C3H2F6O
168.04
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol,hexafluoroisopropanol,hfip,hexafluoro-2-propanol,hexafluoroisopropyl alcohol,bis trifluoromethyl methanol,1,1,1,3,3,3-hexafluoroisopropanol,2h-hexafluoroisopropanol,2-propanol, 1,1,1,3,3,3-hexafluoro,1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O
920-66-1
C3H2F6O
168.04
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol,hexafluoroisopropanol,hfip,hexafluoro-2-propanol,hexafluoroisopropyl alcohol,bis trifluoromethyl methanol,1,1,1,3,3,3-hexafluoroisopropanol,2h-hexafluoroisopropanol,2-propanol, 1,1,1,3,3,3-hexafluoro,1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O
Viscosity 1.65 mPa.s (20°C)
Density 1.5960g/mL
Quantity 100mL
Water 0.02% max. (K.F.)
Packaging Glass bottle
Refractive Index 1.2740 to 1.2770
Melting Point -4.0°C
Specific Gravity 1.596
Residue after Evaporation 0.0005% max.
Color Undesignated
Flash Point >100°C
Boiling Point 59.0°C
CAS Min % 99.75
CAS Max % 100.0
Assay Percent Range 99.8% min. (GC)
Linear Formula (CF3)2CHOH
Solubility Information Solubility in water: 1000g/L (25°C)
Formula Weight 168.04
Physical Form Liquid
Percent Purity 99.8%
Grade For peptide synthesis
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 213-059-4