Alfa Aesar™ 1,1,1,3,3,3-Hexafluoroacetone trideuterate, 99.5% (Isotopic)

Manufacturer: Alfa Aesar™ 4229009

Quantity 10g

 View more versions of this product

Catalog No. AA4229009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109640-39-3
    Formula Weight 226.10 (166.02 Anhydrous)
    MDL Number MFCD00149064
    UN Number UN2552
    Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone trideuterate, Isotopic

    Specifications

    Formula Weight 226.10 (166.02 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    CAS 109640-39-3
    Health Hazard 2 GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 1.71g/mL
    Quantity 10g
    MDL Number MFCD00149064
    UN Number UN2552
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAFLUOROACETONE HYDRATE, LIQUID
    Sensitivity Moisture Sensitive
    Melting Point 21°C
    Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone trideuterate, Isotopic
    Assay 99.5%

    GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit