Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Click to view available options
Quantity:
10g
25g
2160-89-6
C6H4F6O2
222.086
MFCD00040104
MNSWITGNWZSAMC-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoroisopropyl acrylate,hexafluoroisopropyl acrylate,1,1,1,3,3,3-hexafluoropropan-2-yl acrylate,2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester,acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester,hexafluoro-2-propyl acrylate,mnswitgnwzsamc-uhfffaoysa,1h-1-trifluoromethyl trifluoroethyl acrylate,1h-1-trifluoromethyl trifluoroethyl acrylate,1,1,1,3,3,3-hexafluoroisopropyl acrylate
75096
1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
C=CC(=O)OC(C(F)(F)F)C(F)(F)F
2160-89-6
C6H4F6O2
222.086
MFCD00040104
MNSWITGNWZSAMC-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoroisopropyl acrylate,hexafluoroisopropyl acrylate,1,1,1,3,3,3-hexafluoropropan-2-yl acrylate,2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester,acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester,hexafluoro-2-propyl acrylate,mnswitgnwzsamc-uhfffaoysa,1h-1-trifluoromethyl trifluoroethyl acrylate,1h-1-trifluoromethyl trifluoroethyl acrylate,1,1,1,3,3,3-hexafluoroisopropyl acrylate
75096
1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
C=CC(=O)OC(C(F)(F)F)C(F)(F)F
Boiling Point 42°C
Color Yellow
Quantity 25g
UN Number 3272
Formula Weight 222.09
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl Acrylate (stabilized with MEHQ)