Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, 99%, stab.
Click to view available options
Quantity:
1g
5g
25g
Specifications
Quantity | 5g |
Refractive Index | 1.331 |
Density | 1.302 |
Boiling Point | 99°C |
Flash Point | 14°C (57°F) |
UN Number | UN1993 |
Formula Weight | 236.12 |
Percent Purity | 99% |
Chemical Name or Material | 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, stabilized |
Safety and Handling
GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H224-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 221-309-9
TSCA : Yes
Recommended Storage : Ambient temperatures