Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Click to view available options
Quantity:
10g
25g
3063-94-3
C7H6F6O2
236.113
MFCD00040105
FMQPBWHSNCRVQJ-UHFFFAOYSA-N
hexafluoroisopropyl methacrylate,1,1,1,3,3,3-hexafluoroisopropyl methacrylate,2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester,methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester,2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate,hexafluoro-2-propyl methacrylate,1h-1-trifluoromethyl trifluoroethyl methacrylate,1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate,1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate,hfipma
76469
1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
3063-94-3
C7H6F6O2
236.113
MFCD00040105
FMQPBWHSNCRVQJ-UHFFFAOYSA-N
hexafluoroisopropyl methacrylate,1,1,1,3,3,3-hexafluoroisopropyl methacrylate,2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester,methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester,2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate,hexafluoro-2-propyl methacrylate,1h-1-trifluoromethyl trifluoroethyl methacrylate,1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate,1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate,hfipma
76469
1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
Color Colorless
Quantity 10g
UN Number 3272
Formula Weight 236.11
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl Methacrylate (stabilized with MEHQ)

EINECSNumber : (2)-1031

TSCA : Yes