Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,1,1,3,3-Pentafluorobutane, 99.5+%

Manufacturer:  Alfa Aesar™ H337370B

Quantity 1000g

 View more versions of this product

Catalog No. AAH337370B


  Check Availability
Add to cart

Specifications

Description

Applications
Used as foam blowing agent in the production of closed cell rigid insulating foams. It is also used as degreasing solvent in semi-closed and closed degreasing systems. It finds also its use as heat exchange fluid in closed heat transfer systems.

Solubility
Soluble in water (1.7g/L at 21.2C). Miscible with most organic solvents.

Notes
Store in cool location. With adequate ventilation. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

406-58-6
CH3CF2CH2CF3
MFCD03094115
Solkane® 365mfc
C4H5F5
148.08
UN1993
1,1,1,3,3-Pentafluorobutane

Specifications

406-58-6
C4H5F5
−20°C (−4°F)
148.08
UN1993
1,1,1,3,3-Pentafluorobutane
40°C
1000g
CH3CF2CH2CF3
MFCD03094115
Solkane® 365mfc
≥99.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit