Alfa Aesar™ 1,1,1,3,3-Pentafluorobutane, 99.5+%

Manufacturer: Alfa Aesar™ H3373730

Quantity 250g

 View more versions of this product

Catalog No. AAH3373730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 406-58-6
    Molecular Formula C4H5F5
    Linear Formula CH3CF2CH2CF3
    Formula Weight 148.08
    MDL Number MFCD03094115
    UN Number UN1993
    Synonym Solkane® 365mfc
    Chemical Name or Material 1,1,1,3,3-Pentafluorobutane

    Specifications

    Formula Weight 148.08
    TSCA TSCA
    Boiling Point 40°C
    Quantity 250g
    Linear Formula CH3CF2CH2CF3
    MDL Number MFCD03094115
    UN Number UN1993
    Synonym Solkane® 365mfc
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C4H5F5
    Flash Point −20°C (−4°F)
    CAS 406-58-6
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 1,1,1,3,3-Pentafluorobutane
    Assay ≥99.5%

    Description

    Applications
    Used as foam blowing agent in the production of closed cell rigid insulating foams. It is also used as degreasing solvent in semi-closed and closed degreasing systems. It finds also its use as heat exchange fluid in closed heat transfer systems.

    Solubility
    Soluble in water (1.7g/L at 21.2C). Miscible with most organic solvents.

    Notes
    Store in cool location. With adequate ventilation. Store away from oxidizing agent.

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit