Learn More
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%, Thermo Scientific Chemicals
Description
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.
Solubility
Soluble in ethanol, ether, chloroform, and glycerol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Melting Point | 98°C to 100°C (anhydrous) |
Boiling Point | 167°C |
Flash Point | >100°C (212°F) |
Quantity | 25 g |
Beilstein | 878167 |
Merck Index | 14,2129 |
Solubility Information | Soluble in ethanol,ether,chloroform,and glycerol. Insoluble in water. |
Formula Weight | 186.47 (177.46 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | 1,1,1-Trichloro-2-methyl-2-propanol hemihydrate |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 200-317-6 |
RTECSNumber | UC0175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.