Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
6001-64-5
0·5 H2O
186.47
MFCD02179352
HBARVHNVVKZUGS-UHFFFAOYSA-N
c4h7cl3o.1/2h2o,1,1,1-trichloro-2-methyl-2-propanol hemihydrate,chlorobutanol hydrogen ion hydrate
102594540
molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
[HH].CC(C)(C(Cl)(Cl)Cl)O.O
6001-64-5
0·5 H2O
186.47
MFCD02179352
HBARVHNVVKZUGS-UHFFFAOYSA-N
c4h7cl3o.1/2h2o,1,1,1-trichloro-2-methyl-2-propanol hemihydrate,chlorobutanol hydrogen ion hydrate
102594540
molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
[HH].CC(C)(C(Cl)(Cl)Cl)O.O
Melting Point 75°C to 79°C
Boiling Point 173°C to 175°C
Flash Point 100°C
Packaging Plastic bottle
Quantity 100g
Color White
Water 4.5 to 5.5% (K.F.)
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula 0·5H2O
Solubility Information Solubility in water: 7.7g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,glycerol
Formula Weight 186.47
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 97.5 to 102.5% (ex Cl)
Chemical Name or Material 1, 1, 1-Trichloro-2-methyl-2-propanol hemihydrate, 98%
Show More Show Less

GHS P Statement Harmful if swallowed.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

TSCA : TSCA