Alfa Aesar™ 1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 95%

Manufacturer: Alfa Aesar™ A1011206

Quantity 5g

 View more versions of this product

Catalog No. AAA1011206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22767-90-4
    Molecular Formula C8H11F3O2
    Formula Weight 196.17
    MDL Number MFCD00040841
    UN Number UN1224
    Beilstein 1708830
    Synonym Pivaloyltrifluoroacetone; PTA
    Chemical Name or Material 1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione

    Specifications

    Formula Weight 196.17
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 22767-90-4
    UN Number UN1224
    Molecular Formula C8H11F3O2
    Synonym Pivaloyltrifluoroacetone; PTA
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    TSCA TSCA
    MDL Number MFCD00040841
    Beilstein 1708830
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    Flash Point 34°C (93°F)
    Boiling Point 68° to 71°C (6mmHg)
    Quantity 5g
    Chemical Name or Material 1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione
    Assay 95%

    Description

    Applications
    Useful for solvent extraction and GC separation of metals.

    Notes
    Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit