Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Trifluoroacetone, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
421-50-1
C3H3F3O
112.05
FHUDAMLDXFJHJE-UHFFFAOYSA-N
1,1,1-trifluoroacetone,trifluoroacetone,1,1,1-trifluoro-2-propanone,methyl trifluoromethyl ketone,2-propanone, 1,1,1-trifluoro,trifluoromethyl methyl ketone,3,3,3-trifluoroacetone,1,1,1,-trifluoroacetone,trifluoroketone,trifluoracetone
9871
1,1,1-trifluoropropan-2-one
CC(=O)C(F)(F)F
421-50-1
C3H3F3O
112.05
FHUDAMLDXFJHJE-UHFFFAOYSA-N
1,1,1-trifluoroacetone,trifluoroacetone,1,1,1-trifluoro-2-propanone,methyl trifluoromethyl ketone,2-propanone, 1,1,1-trifluoro,trifluoromethyl methyl ketone,3,3,3-trifluoroacetone,1,1,1,-trifluoroacetone,trifluoroketone,trifluoracetone
9871
1,1,1-trifluoropropan-2-one
CC(=O)C(F)(F)F
Specific Gravity 1.25
Viscosity 3.28 mPa.s (15°C)
Infrared Spectrum Authentic
Melting Point -78.0°C
Color Colorless
Flash Point −30°C
Packaging Glass bottle
Boiling Point 22.0°C
Refractive Index 1.3000 (20°C, 589nm) max.
Quantity 500g
Density 1.2500g/mL
Assay Percent Range 98+%
Linear Formula CF3COCH3
Beilstein 01,II,717
Fieser 13,323; 15,212
Solubility Information Solubility in water: miscible
Formula Weight 112.05
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material 1, 1, 1-Trifluoroacetone
Show More Show Less

GHS P Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. Extremely flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger