Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Tris(hydroxymethyl)ethane 97%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Plastic bottle
77-85-0
C5H12O3
120.15
MFCD00004687
QXJQHYBHAIHNGG-UHFFFAOYSA-N
trimethylolethane,1,1,1-tris hydroxymethyl ethane,trimet,methriol,metriol,pentaglycerine,pentaglycerol,methyltrimethanolmethane,2-hydroxymethyl-2-methylpropane-1,3-diol,tris hydroxymethyl ethane
6502
2-(hydroxymethyl)-2-methylpropane-1,3-diol
CC(CO)(CO)CO
77-85-0
C5H12O3
120.15
MFCD00004687
QXJQHYBHAIHNGG-UHFFFAOYSA-N
trimethylolethane,1,1,1-tris hydroxymethyl ethane,trimet,methriol,metriol,pentaglycerine,pentaglycerol,methyltrimethanolmethane,2-hydroxymethyl-2-methylpropane-1,3-diol,tris hydroxymethyl ethane
6502
2-(hydroxymethyl)-2-methylpropane-1,3-diol
CC(CO)(CO)CO
Flash Point 150°C
Color White
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 200.0°C to 203.0°C
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C(CH2OH)3
Beilstein 01, 520
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, insoluble in ether, benzene
Formula Weight 120.15
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity 97%
Chemical Name or Material 1, 1, 1-Tris(hydroxymethyl)ethane, 97%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 201-063-9

RTECSNumber : TY7137500

TSCA : TSCA