Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrabromoethane, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Glass bottle
79-27-6
C2H2Br4
345.64
MFCD00000133
QXSZNDIIPUOQMB-UHFFFAOYSA-N
acetylene tetrabromide,muthmann's liquid,ethane, 1,1,2,2-tetrabromo,tetrabromoacetylene,sym-tetrabromoethane,s-tetrabromoethane,1,1,2,2-tetrabromaethan,1,1,2,2-tetrabromoetano,tbe,unii-v00gkc584o
6588
1,1,2,2-tetrabromoethane
C(C(Br)Br)(Br)Br
79-27-6
C2H2Br4
345.64
MFCD00000133
QXSZNDIIPUOQMB-UHFFFAOYSA-N
acetylene tetrabromide,muthmann's liquid,ethane, 1,1,2,2-tetrabromo,tetrabromoacetylene,sym-tetrabromoethane,s-tetrabromoethane,1,1,2,2-tetrabromaethan,1,1,2,2-tetrabromoetano,tbe,unii-v00gkc584o
6588
1,1,2,2-tetrabromoethane
C(C(Br)Br)(Br)Br
Refractive Index 1.6355 to 1.638
Melting Point 1°C
Boiling Point 244°C
Specific Gravity 2.96
Infrared Spectrum Authentic
Quantity 250g
Color Undesignated
Packaging Glass bottle
Density 2.9600g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula Br2CHCHBr2
Beilstein 01, 94
Merck Index 15, 9330
Solubility Information Solubility in water: 0.63g/L (20°C). Other solubilities: soluble in acetone,benzene,miscible with ethanol,ether,chloroform,aniline,and glacial acetic acid
Formula Weight 345.64
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1,1,2,2-Tetrabromoethane
Show More Show Less

GHS H Statement:
Fatal if inhaled.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-191-5