Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrabromoethane, Practical, 95%, Spectrum™

Click to view available options
Quantity:
500 g
5 kg
150 kg
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
79-27-6
79-27-6
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 1
Percent Purity 0.95
Grade Practical

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.