Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane 98.5%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
1L
2.5L
Packaging:
Glass bottle
79-34-5
C2H2Cl4
167.838
MFCD00000848
QPFMBZIOSGYJDE-UHFFFAOYSA-N
s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan
6591
CHEBI:36026
1,1,2,2-tetrachloroethane
C(C(Cl)Cl)(Cl)Cl
79-34-5
C2H2Cl4
167.838
MFCD00000848
QPFMBZIOSGYJDE-UHFFFAOYSA-N
s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan
6591
CHEBI:36026
1,1,2,2-tetrachloroethane
C(C(Cl)Cl)(Cl)Cl
Density 1.5800g/mL
Specific Gravity 1.58
Color Colorless
Melting Point -43.0°C
Quantity 25mL
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 147.0°C
Refractive Index 1.4930 to 1.4950
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98.5%
Linear Formula Cl2CHCHCl2
Beilstein 01, 86
Merck Index 15, 9334
Solubility Information Solubility in water: 0.3 g/100mL (25°C). Other solubilities: miscible with methanol, ethanol, benzene, ether,, petroleum ether, carbon tetrachloride, chloroform,, carbon disulfide, dimethylformamide and oils, soluble in acetone
Formula Weight 167.85
Physical Form Liquid
Percent Purity 98.5%
Chemical Name or Material 1,1,2,2-Tetrachloroethane, 98.5%
Show More Show Less

GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-197-8

RTECSNumber : KI8575000

TSCA : TSCA