Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane, 98%, Spectrum™

Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
79-34-5
C2H2Cl4
167.84
QPFMBZIOSGYJDE-UHFFFAOYSA-N
1,1,2,2-tetrachloroethane
ClC(Cl)C(Cl)Cl
79-34-5
C2H2Cl4
167.84
QPFMBZIOSGYJDE-UHFFFAOYSA-N
1,1,2,2-tetrachloroethane
ClC(Cl)C(Cl)Cl

T1018, 79-34-5, C2H2Cl4

Spectrum™ 1,1,2,2-Tetrachloroethane , also known as Acetylene tetrachloride, is a chlorinated derivative of ethane with a very high solvent power. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 100%
UN Number UN1702
Percent Purity 98%