Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane-d2 99 atom% D, ACROS Organics™

Click to view available options
Quantity:
5mL
20mL
Packaging:
Glass bottle
33685-54-0
C2H2Cl4
169.85
MFCD00037672
QPFMBZIOSGYJDE-QDNHWIQGSA-N
1,1,2,2-tetrachloroethane-d2,1,2-dideutero-1,1,2,2-tetrachloroethane,tetrachloro 2 h? ethane,1,1,2,2-tetrachloroethane-d2 tce-d2,ethane-d2, 1,1,2,2-tetrachloro,ethane-1,2-d2, 1,1,2,2-tetrachloro,1,1,2,2-tetrachloro-1,2-2h2 ethane,de87d,1,1,2,2-tetrachloroethane-d2 >99.50 atom % d,1,1,2,2-tetrachloroethane-d2, >=99.5 atom % d
118531
1,1,2,2-tetrachloro-1,2-dideuterioethane
C(C(Cl)Cl)(Cl)Cl
33685-54-0
C2H2Cl4
169.85
MFCD00037672
QPFMBZIOSGYJDE-QDNHWIQGSA-N
1,1,2,2-tetrachloroethane-d2,1,2-dideutero-1,1,2,2-tetrachloroethane,tetrachloro 2 h? ethane,1,1,2,2-tetrachloroethane-d2 tce-d2,ethane-d2, 1,1,2,2-tetrachloro,ethane-1,2-d2, 1,1,2,2-tetrachloro,1,1,2,2-tetrachloro-1,2-2h2 ethane,de87d,1,1,2,2-tetrachloroethane-d2 >99.50 atom % d,1,1,2,2-tetrachloroethane-d2, >=99.5 atom % d
118531
1,1,2,2-tetrachloro-1,2-dideuterioethane
C(C(Cl)Cl)(Cl)Cl
Color Colorless
Quantity 5mL
Density 1.6100g/mL
Deuterium (D) 98.5% min.
Packaging Glass bottle
Boiling Point 146.0°C
Linear Formula (Cl2CD)2
Beilstein 01, III, 164
Refractive Index 1.4924
Formula Weight 169.86
Physical Form Liquid
Grade NMR
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2, 99 atom % D
Show More Show Less

GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 251-634-1

TSCA : TSCA