Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1,1,2,2-Tetrafluoroethoxy)benzene, 98%

Click to view available options
Quantity:
1g
5g
25g
350-57-2
C8H6F4O
194.129
MFCD00042102
GRDIVJPQARIBNZ-UHFFFAOYSA-N
1,1,2,2-tetrafluoroethoxy benzene,tetrafluoroethoxy benzene,fentalene-14,benzene, 1,1,2,2-tetrafluoroethoxy,1,1,2,2-tetrafluoro-1-phenoxyethane,3-06-00-00598 beilstein handbook reference,benzene, tetrafluoroethoxy,ether, phenyl tetrafluoroethoxy,tetrafluoroethyl ether of phenol,4-1,1,2,2-tetrafluoroethoxy benzene
67692
1,1,2,2-tetrafluoroethoxybenzene
C1=CC=C(C=C1)OC(C(F)F)(F)F
350-57-2
C8H6F4O
194.129
MFCD00042102
GRDIVJPQARIBNZ-UHFFFAOYSA-N
1,1,2,2-tetrafluoroethoxy benzene,tetrafluoroethoxy benzene,fentalene-14,benzene, 1,1,2,2-tetrafluoroethoxy,1,1,2,2-tetrafluoro-1-phenoxyethane,3-06-00-00598 beilstein handbook reference,benzene, tetrafluoroethoxy,ether, phenyl tetrafluoroethoxy,tetrafluoroethyl ether of phenol,4-1,1,2,2-tetrafluoroethoxy benzene
67692
1,1,2,2-tetrafluoroethoxybenzene
C1=CC=C(C=C1)OC(C(F)F)(F)F
Flash Point 46°C (115°F)
Boiling Point 152°C to 153°C
Quantity 5g
UN Number UN1993
Formula Weight 194.13
Percent Purity 98%
Chemical Name or Material (1,1,2,2-Tetrafluoroethoxy)benzene

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : DC0340000