Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2-Trimethyl-1H-benzo[e]indole, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
41532-84-7
C15H15N
209.292
MFCD00082627
WJZSZXCWMATYFX-UHFFFAOYSA-N
1,1,2-trimethyl-1h-benzo e indole,1,1,2-trimethyl-1h-benz e indole,1,1,2-trimethylbenz e indole,2,3,3-trimethyl-4,5-benzo-3h-indole,2,3,3-trimethylbenzoindolenine,1,1,2-trimethylbenzo e indole,1h-benz e indole, 1,1,2-trimethyl,2,3,3-trimethyl-1h-benz e indolenine,1,1,2-trimethylhydrobenzo e indole
170530
1,1,2-trimethylbenzo[e]indole
CC1=NC2=C(C1(C)C)C3=CC=CC=C3C=C2
41532-84-7
C15H15N
209.292
MFCD00082627
WJZSZXCWMATYFX-UHFFFAOYSA-N
1,1,2-trimethyl-1h-benzo e indole,1,1,2-trimethyl-1h-benz e indole,1,1,2-trimethylbenz e indole,2,3,3-trimethyl-4,5-benzo-3h-indole,2,3,3-trimethylbenzoindolenine,1,1,2-trimethylbenzo e indole,1h-benz e indole, 1,1,2-trimethyl,2,3,3-trimethyl-1h-benz e indolenine,1,1,2-trimethylhydrobenzo e indole
170530
1,1,2-trimethylbenzo[e]indole
CC1=NC2=C(C1(C)C)C3=CC=CC=C3C=C2
Applications
Reactant used for preparation of fluorescent probes for imaging of tumors in vivo, hypoxia-sensitive fluorescent probes for in vivo real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Quantity 1g
Melting Point 114°C to 119°C
Beilstein 153709
Formula Weight 209.29
Percent Purity 97%
Chemical Name or Material 1,1,2-Trimethyl-1H-benzo[e]indole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA