Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3,3-Tetramethyl-2-thiourea 98%, Acros Organics

Click to view available options
Quantity:
25g
2782-91-4
C5H12N2S
132.225
MFCD00008324
MNOILHPDHOHILI-UHFFFAOYSA-N
tetramethylthiourea,thiourea, tetramethyl,basthioryl,1,1,3,3-tetramethyl-2-thiourea,tmtu,n,n,n',n'-tetramethylthiourea,urea, thio-, tetramethyl,tetramethyl-2-thiourea,urea, 1,1,3,3-tetramethyl-2-thio,thiourea, n,n,n',n'-tetramethyl
17725
1,1,3,3-tetramethylthiourea
CN(C)C(=S)N(C)C
2782-91-4
C5H12N2S
132.225
MFCD00008324
MNOILHPDHOHILI-UHFFFAOYSA-N
tetramethylthiourea,thiourea, tetramethyl,basthioryl,1,1,3,3-tetramethyl-2-thiourea,tmtu,n,n,n',n'-tetramethylthiourea,urea, thio-, tetramethyl,tetramethyl-2-thiourea,urea, 1,1,3,3-tetramethyl-2-thio,thiourea, n,n,n',n'-tetramethyl
17725
1,1,3,3-tetramethylthiourea
CN(C)C(=S)N(C)C
Quantity 25g
Melting Point 74°C to 77°C
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2NCSN(CH3)2
Beilstein 04, I, 336
Fieser 01,1145; 07,359
Formula Weight 132.22
Physical Form Crystalline Chunks
Percent Purity ≥97.5%
Chemical Name or Material 1,1,3,3-Tetramethyl-2-thiourea, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.