Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,1,3,3-Tetramethyldisiloxane, 97%, ACROS Organics™
Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
Description
Reduction reagent
Specifications
Specific Gravity | 0.76 |
Refractive Index | 1.3690 to 1.3710 |
Quantity | 25mL |
Density | 0.7600g/mL |
Flash Point | −20°C |
Viscosity | 0.5 mPa.s (25°C) |
Color | Colorless |
Melting Point | -78.0°C |
Boiling Point | 70.0°C to 71.0°C |
Infrared Spectrum | Authentic |
Show More |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai
GHS Signal Word: Danger
EINECSNumber : 221-906-4