Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
306935-39-7
C11H7F3O4
260.168
MFCD01570563
FDLJUCOQPAGQNY-UHFFFAOYSA-N
1-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1-2h-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 3-3,4-methylenedioxy benzoyl-1,1,1-trifluoroacetone, 1-benzo d 1,3 dioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, maybridge1_005379, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dion, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1,3-butanedione,1-1,3-benzodioxol-5-yl-4,4,4-trifluoro, 1-1,3-benzodioxol-5-yl-4,4,4-tri-fluorobutane-1,3-dione, 1-1,3-benzodioxole-5-yl-4,4,4-trifluorobutane-1,3-dione
2775368
1-(1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
C1OC2=C(O1)C=C(C=C2)C(=O)CC(=O)C(F)(F)F
306935-39-7
C11H7F3O4
260.168
MFCD01570563
FDLJUCOQPAGQNY-UHFFFAOYSA-N
1-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1-2h-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 3-3,4-methylenedioxy benzoyl-1,1,1-trifluoroacetone, 1-benzo d 1,3 dioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, maybridge1_005379, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dion, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1,3-butanedione,1-1,3-benzodioxol-5-yl-4,4,4-trifluoro, 1-1,3-benzodioxol-5-yl-4,4,4-tri-fluorobutane-1,3-dione, 1-1,3-benzodioxole-5-yl-4,4,4-trifluorobutane-1,3-dione
2775368
1-(1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
C1OC2=C(O1)C=C(C=C2)C(=O)CC(=O)C(F)(F)F
Quantity 1g
Packaging Amber Glass Bottle
CAS Min % 97.0
Assay Percent Range 97%
Formula Weight 260.17
Percent Purity 97%
Chemical Name or Material 1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.