Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,1,3-Trimethoxypropane, 98%

Click to view available options
Quantity:
5g
25g
100g
14315-97-0
C6H14O3
134.175
MFCD00008490
FKZYYYDRLJCHGL-UHFFFAOYSA-N
propane, 1,1,3-trimethoxy,propane,1,1,3-trimethoxy,acmc-1brb6,1,1,3-trimethoxy-propane,1,1,3-trimethoxypropane,beta-methoxypropionaldehyde dimethyl acetal
84348
1,1,3-trimethoxypropane
COCCC(OC)OC
14315-97-0
C6H14O3
134.175
MFCD00008490
FKZYYYDRLJCHGL-UHFFFAOYSA-N
propane, 1,1,3-trimethoxy,propane,1,1,3-trimethoxy,acmc-1brb6,1,1,3-trimethoxy-propane,1,1,3-trimethoxypropane,beta-methoxypropionaldehyde dimethyl acetal
84348
1,1,3-trimethoxypropane
COCCC(OC)OC
Flash Point 40°C (105°F)
Boiling Point 45°C to 46°C (17mmHg)
Quantity 100g
UN Number UN3271
Formula Weight 134.17
Percent Purity 98%
Chemical Name or Material 1,1,3-Trimethoxypropane

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

RTECSNumber : TZ9560000