Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,1,3-Triphenylpropargyl alcohol, 97%

Manufacturer:  Alfa Aesar™ H5346114

Quantity 25g

 View more versions of this product

Catalog No. AAH5346114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1522-13-0
284.36
1,1,3-Triphenyl-2-propyn-1-ol
C21H16O
MFCD00004449
1,1,3-Triphenylpropargyl alcohol

Specifications

1522-13-0
284.36
1,1,3-Triphenyl-2-propyn-1-ol
25g
97%
C21H16O
MFCD00004449
80° to 82°C
1,1,3-Triphenylpropargyl alcohol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit