Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-(4,6-Dihydroxy-1,3-phenylene)bisethanone, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
2161-85-5
C10H10O4
194.186
MFCD00017655
GEYCQLIOGQPPFM-UHFFFAOYSA-N
4,6-diacetylresorcinol,resodiacetophenone,1,1'-4,6-dihydroxy-1,3-phenylene bisethanone,ethanone, 1,1'-4,6-dihydroxy-1,3-phenylene bis,1-5-acetyl-2,4-dihydroxyphenyl ethanone,1,1'-4,6-dihydroxy-1,3-phenylene diethanone,1,3-diacetyl-4,6-dihydroxybenzene,ethanone,1,1'-4,6-dihydroxy-1,3-phenylene bis,1-5-acetyl-2,4-dihydroxyphenyl ethan-1-one,1,1'-4,6-dihydroxy-1,3-phenylene-bis-ethanone
75097
1-(5-acetyl-2,4-dihydroxyphenyl)ethanone
CC(=O)C1=CC(=C(C=C1O)O)C(=O)C
2161-85-5
C10H10O4
194.186
MFCD00017655
GEYCQLIOGQPPFM-UHFFFAOYSA-N
4,6-diacetylresorcinol,resodiacetophenone,1,1'-4,6-dihydroxy-1,3-phenylene bisethanone,ethanone, 1,1'-4,6-dihydroxy-1,3-phenylene bis,1-5-acetyl-2,4-dihydroxyphenyl ethanone,1,1'-4,6-dihydroxy-1,3-phenylene diethanone,1,3-diacetyl-4,6-dihydroxybenzene,ethanone,1,1'-4,6-dihydroxy-1,3-phenylene bis,1-5-acetyl-2,4-dihydroxyphenyl ethan-1-one,1,1'-4,6-dihydroxy-1,3-phenylene-bis-ethanone
75097
1-(5-acetyl-2,4-dihydroxyphenyl)ethanone
CC(=O)C1=CC(=C(C=C1O)O)C(=O)C
Infrared Spectrum Authentic
Quantity 5g
Melting Point 178°C to 185°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (HO)C6H(COCH3)
Formula Weight 194.19
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 1,1'-(4,6-Dihydroxy-1,3-phenylene)bisethanone
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.