Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,4-Trimethylcyclohexane 98.0+%, TCI America™

Click to view available options
Quantity:
1mL
7094-27-1
C9H18
126.243
MFCD00058950
UIWORXHEVNIOJG-UHFFFAOYSA-N
35365
1,1,4-trimethylcyclohexane
CC1CCC(CC1)(C)C
7094-27-1
C9H18
126.243
MFCD00058950
UIWORXHEVNIOJG-UHFFFAOYSA-N
35365
1,1,4-trimethylcyclohexane
CC1CCC(CC1)(C)C
Color Colorless
Quantity 1mL
Boiling Point 135°C
UN Number 3295
Formula Weight 126.24
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,1,4-Trimethylcyclohexane