Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(1-Adamantyl)ethylamine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
Solubility
Freely soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
Solubility
Freely soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Melting Point | 375°C |
Quantity | 5 g |
Merck Index | 148224 |
Solubility Information | Freely soluble in water. |
Formula Weight | 215.77 |
Percent Purity | 98% |
Chemical Name or Material | 1-(1-Adamantyl)ethylamine hydrochloride |
Safety and Handling
Hazard Category | Acute toxicity (category 4) |
Hazard Statement | Harmful if swallowed |
RTECSNumber | AU4717000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction