Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-1,1'-Bi(2-naphthol), 99%

Manufacturer:  Alfa Aesar™ A1118214

Quantity 25g

 View more versions of this product

Catalog No. AAA1118214


  Check Availability
Add to cart

Specifications

Description

Applications
(+/-)-1,1 ft.-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

Solubility
Soluble in dioxane (50 mg/mL). Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Refrigerated (2-8°C)
Description & Specifications

Chemical Identifiers

602-09-5
286.33
UN2811
14,1226
(±)-1,1'-Bi (2-naphthol)
C20H14O2
MFCD00004068
997518
(+/-)-Binaphthalene-2,2'-diol; (+/-)-BINOL

Specifications

602-09-5
C20H14O2
MFCD00004068
997518
14,1226
25g
99%
Characteristic
286.33
UN2811
215° to 218°C
(+/-)-Binaphthalene-2,2'-diol; (+/-)-BINOL
(±)-1,1'-Bi (2-naphthol)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DU3106000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit