Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bi-2-naphthol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
602-09-5
C20H14O2
286.32
MFCD00004068
PPTXVXKCQZKFBN-UHFFFAOYSA-N
1,1'-bi-2-naphthol,s---1,1'-bi-2-naphthol,r-+-1,1'-bi-2-naphthol,1,1'-binaphthalene-2,2'-diol,binol,1,1'-binaphthyl-2,2'-diol,2,2'-dinaphthol,beta-binaphthol,r-1,1'-bi-2-naphthol,s-1,1'-binaphthalene-2,2'-diol
11762
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
602-09-5
C20H14O2
286.32
MFCD00004068
PPTXVXKCQZKFBN-UHFFFAOYSA-N
1,1'-bi-2-naphthol,s---1,1'-bi-2-naphthol,r-+-1,1'-bi-2-naphthol,1,1'-binaphthalene-2,2'-diol,binol,1,1'-binaphthyl-2,2'-diol,2,2'-dinaphthol,beta-binaphthol,r-1,1'-bi-2-naphthol,s-1,1'-binaphthalene-2,2'-diol
11762
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 215°C to 218°C
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC10H6C10H6OH
Beilstein 06, 1051
Fieser 15,26; 16,24
Merck Index 15, 1230
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acetone and ethanol
Formula Weight 286.32
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 1, 1'-Bi-2-naphthol, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Toxic if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-014-0

RTECSNumber : DU3106000

TSCA : TSCA