Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[1,1«-Biphenyl]-3-ylboronic acid, 97%, Acros Organics

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
5122-95-2
C12H11BO2
198.028
GOXICVKOZJFRMB-UHFFFAOYSA-N
3-biphenylboronic acid,biphenyl-3-boronic acid,1,1'-biphenyl-3-ylboronic acid,biphenyl-3-ylboronic acid,3-phenylphenyl boranediol,3-phenylphenyl boronic acid,3-biphenyl boronic acid,3-phenylbenzeneboronic acid,3-biphenylylboronic acid,1,1'-biphenyl-3-yl boronic acid
2734315
(3-phenylphenyl)boronic acid
B(C1=CC(=CC=C1)C2=CC=CC=C2)(O)O
5122-95-2
C12H11BO2
198.028
GOXICVKOZJFRMB-UHFFFAOYSA-N
3-biphenylboronic acid,biphenyl-3-boronic acid,1,1'-biphenyl-3-ylboronic acid,biphenyl-3-ylboronic acid,3-phenylphenyl boranediol,3-phenylphenyl boronic acid,3-biphenyl boronic acid,3-phenylbenzeneboronic acid,3-biphenylylboronic acid,1,1'-biphenyl-3-yl boronic acid
2734315
(3-phenylphenyl)boronic acid
B(C1=CC(=CC=C1)C2=CC=CC=C2)(O)O
Color Beige
Quantity 250mg
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 198.03
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Biphenyl-3-boronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.