Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
250mg
Packaging:
Glass bottle
849773-97-3
C38H60BF4FeP2Rh-
824.4
MFSWTFGIDMKBOY-VXMPMTHESA-N
1,1 inverted exclamation marka-bis 2r,5r-2,5-diisopropylphospholano ferrocene cyclooctadiene rhodium i tetrafluoroborate
71310759
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-cyclopenta-2,4-dien-1-yl-2,5-di(propan-2-yl)phospholane;iron(2+);rhodium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1CCC(P1[C-]2C=CC=C2)C(C)C.CC(C)C1CCC(P1[C-]2C=CC=C2)C(C)C.C1CC=CCCC=C1.[Fe+2].[Rh]
849773-97-3
C38H60BF4FeP2Rh-
824.4
MFSWTFGIDMKBOY-VXMPMTHESA-N
1,1 inverted exclamation marka-bis 2r,5r-2,5-diisopropylphospholano ferrocene cyclooctadiene rhodium i tetrafluoroborate
71310759
(1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-cyclopenta-2,4-dien-1-yl-2,5-di(propan-2-yl)phospholane;iron(2+);rhodium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)C1CCC(P1[C-]2C=CC=C2)C(C)C.CC(C)C1CCC(P1[C-]2C=CC=C2)C(C)C.C1CC=CCCC=C1.[Fe+2].[Rh]
Color Yellow to Brown
Melting Point 235.0°C to 238.0°C
Packaging Glass bottle
Quantity 100mg
NMR 96.0% min.
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: insoluble.
Formula Weight 824.39
Physical Form Crystalline Powder or Crystals
Percent Purity 97%
Chemical Name or Material 1,1′-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less