Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ferrocene, 97+%, Thermo Scientific™

Click to view available options
Quantity:
250mg
540475-45-4
C24H48FeP2
454.44
MFCD08705241,MFCD08705241
JBWUTRKMUDYCHS-UHFFFAOYNA-N
-1,1'-bis 2r,5r-2,5-dimethylphospholano ferrocene
132984027
λ²-iron(2+) bis(1-cyclopentyl-2,5-dimethylphospholane) dimethanide
[CH3-].[CH3-].[Fe++].CC1CCC(C)P1C1CCCC1.CC1CCC(C)P1C1CCCC1
540475-45-4
C24H48FeP2
454.44
MFCD08705241,MFCD08705241
JBWUTRKMUDYCHS-UHFFFAOYNA-N
-1,1'-bis 2r,5r-2,5-dimethylphospholano ferrocene
132984027
λ²-iron(2+) bis(1-cyclopentyl-2,5-dimethylphospholane) dimethanide
[CH3-].[CH3-].[Fe++].CC1CCC(C)P1C1CCCC1.CC1CCC(C)P1C1CCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
Melting Point 99°C to 102°C
Quantity 250mg
Solubility Information Insoluble in water.
Sensitivity Air Sensitive
Formula Weight 414.28
Percent Purity ≥97%
Chemical Name or Material 1,1'-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ferrocene

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen