Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ferrocene, 97+%, Thermo Scientific™

Click to view available options
Quantity:
250mg
162412-87-5
C22H32FeP2
414.291
MFCD09265089
QXXSYUGLJIMLFH-VUTIHBPYSA-N
(S,S)-Methyl-Ferrocelane
71310453
(2S,5S)-1-cyclopenta-2,4-dien-1-yl-2,5-dimethylphospholane;iron(2+)
CC1CCC(P1[C-]2C=CC=C2)C.CC1CCC(P1[C-]2C=CC=C2)C.[Fe+2]
162412-87-5
C22H32FeP2
414.291
MFCD09265089
QXXSYUGLJIMLFH-VUTIHBPYSA-N
(S,S)-Methyl-Ferrocelane
71310453
(2S,5S)-1-cyclopenta-2,4-dien-1-yl-2,5-dimethylphospholane;iron(2+)
CC1CCC(P1[C-]2C=CC=C2)C.CC1CCC(P1[C-]2C=CC=C2)C.[Fe+2]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep away from strong oxidizing agents.
Melting Point 88°C to 99°C
Quantity 250mg
Solubility Information Insoluble in water.
Sensitivity Air Sensitive
Formula Weight 414.28
Optical Rotation −331° (c=1 in Chloroform)
Percent Purity ≥97%
Chemical Name or Material 1,1'-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ferrocene

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen