Alfa Aesar™ 1,1'-Bis(dichlorophosphino)ferrocene, 98%

Manufacturer: Alfa Aesar™ H2729303

Quantity 1g

 View more versions of this product

Catalog No. AAH2729303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142691-70-1
    Molecular Formula C10H8Cl4FeP2
    Formula Weight 387.78
    MDL Number MFCD01630854
    UN Number UN3261
    Chemical Name or Material 1,1'-Bis(dichlorophosphino)ferrocene

    Specifications

    Formula Weight 387.78
    UN Number UN3261
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C10H8Cl4FeP2
    MDL Number MFCD01630854
    CAS 142691-70-1
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 1,1'-Bis(dichlorophosphino)ferrocene
    Assay 98%

    Description

    Applications
    It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.

    Solubility
    Reacts with water.

    Notes
    Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit