Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(diisopropylphosphino)ferrocene, 98%, ACROS Organics™

Click to view available options
Quantity:
2.5g
Packaging:
Glass bottle
97239-80-0
C22H36FeP2
418.32
MFCD02684559
97239-80-0
C22H36FeP2
418.32
MFCD02684559
Quantity 2.5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Orange
Melting Point 53.0°C to 57.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble.
Formula Weight 418.32
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1,1′-Bis(diisopropylphosphino)ferrocene, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA