Alfa Aesar™ 1,1-Bis(diphenylphosphino)ethylene, 98%

Manufacturer: Alfa Aesar™ H5586403

Quantity 1g

 View more versions of this product

Catalog No. AAH5586403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 84494-89-3
    Linear Formula [(C6H5)2P]2C=CH2
    Formula Weight 396.41
    MDL Number MFCD00191809
    Chemical Name or Material 1,1-Bis(diphenylphosphino)ethylene

    Specifications

    Formula Weight 396.41
    Melting Point 110° to 111°C
    Physical Form Solid
    CAS 84494-89-3
    Quantity 1g
    Linear Formula [(C6H5)2P]2C=CH2
    MDL Number MFCD00191809
    Chemical Name or Material 1,1-Bis(diphenylphosphino)ethylene
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit