Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(diphenylphosphino)ferrocene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
12150-46-8
C34H28FeP2
MFCD00001422
12150-46-8
C34H28FeP2
MFCD00001422
Quantity 25g
Color Orange
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 3490
Formula Weight 553.99
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 1, 1'-Bis(diphenylphosphino)ferrocene
Show More Show Less

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger