Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
75-34-3,67-56-1
C2H4Cl2
98.954
MFCD00792899
SCYULBFZEHDVBN-UHFFFAOYSA-N
ethylidene chloride,ethane, 1,1-dichloro,ethylidene dichloride,dichloroethane,1,1-dichlorethane,aethylidenchlorid,1,1-dicloroetano,1,1-ethylidene dichloride,dichloromethylmethane,rcra waste number u076
6365
CHEBI:81599
1,1-dichloroethane
CC(Cl)Cl
75-34-3,67-56-1
C2H4Cl2
98.954
MFCD00792899
SCYULBFZEHDVBN-UHFFFAOYSA-N
ethylidene chloride,ethane, 1,1-dichloro,ethylidene dichloride,dichloroethane,1,1-dichlorethane,aethylidenchlorid,1,1-dicloroetano,1,1-ethylidene dichloride,dichloromethylmethane,rcra waste number u076
6365
CHEBI:81599
1,1-dichloroethane
CC(Cl)Cl
Quantity 1g
Packaging Glass bottle
Color Brown-Red to Orange
Infrared Spectrum Authentic
Elemental Analysis 50 to 53% (C)
Formula Weight 816.22
Physical Form Crystalline Powder
Chemical Name or Material 1, 1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning