Alfa Aesar™ [1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonylmolybdenum, 98%

Manufacturer: Alfa Aesar™ 4428106

Quantity 5g

 View more versions of this product

Catalog No. AA4428106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67292-28-8
    Molecular Formula C38H28FeMoO4P2
    Formula Weight 762.37
    MDL Number MFCD03427159
    Chemical Name or Material [1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylmolybdenum

    Specifications

    Formula Weight 762.37
    Physical Form Powder
    CAS 67292-28-8
    Molecular Formula C38H28FeMoO4P2
    MDL Number MFCD03427159
    Melting Point 256°C (dec.)
    Quantity 5g
    Chemical Name or Material [1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylmolybdenum
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit