Alfa Aesar™ 1,1-Bis(methylthio)-2-nitroethylene, 99%

Manufacturer: Alfa Aesar™ A1178922

Quantity 100g

 View more versions of this product

Catalog No. AAA1178922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13623-94-4
    Molecular Formula C4H7NO2S2
    Formula Weight 165.23
    MDL Number MFCD00010443
    Beilstein 1524538
    Synonym 1,1-Bis(methylmercapto)-2-nitroethylene
    Chemical Name or Material 1,1-Bis (methylthio)-2-nitroethylene

    Specifications

    Formula Weight 165.23
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00010443
    Molecular Formula C4H7NO2S2
    CAS 13623-94-4
    Synonym 1,1-Bis(methylmercapto)-2-nitroethylene
    Melting Point 124° to 127°C
    Quantity 100g
    Health Hazard 1 Warning
    Beilstein 1524538
    Odor Unpleasant
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 1,1-Bis (methylthio)-2-nitroethylene
    Assay 99%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit