Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis(trichlorosilylmethyl)ethylene, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
78948-04-6
C4H6Cl6Si2
322.962
MFCD00239510
LQEUNVMRBBCABI-UHFFFAOYSA-N
1,1-bis trichlorosilylmethyl ethylene,trichloro-2-trichlorosilylmethyl prop-2-enyl silane,trichloro 2-trichlorosilyl methyl prop-2-en-1-yl silane
2733840
trichloro-[2-(trichlorosilylmethyl)prop-2-enyl]silane
C=C(C[Si](Cl)(Cl)Cl)C[Si](Cl)(Cl)Cl
78948-04-6
C4H6Cl6Si2
322.962
MFCD00239510
LQEUNVMRBBCABI-UHFFFAOYSA-N
1,1-bis trichlorosilylmethyl ethylene,trichloro-2-trichlorosilylmethyl prop-2-enyl silane,trichloro 2-trichlorosilyl methyl prop-2-en-1-yl silane
2733840
trichloro-[2-(trichlorosilylmethyl)prop-2-enyl]silane
C=C(C[Si](Cl)(Cl)Cl)C[Si](Cl)(Cl)Cl
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Colorless
Refractive Index 1.4870 to 1.4890 (20°C, 589nm)
Linear Formula Cl3SiCH2C(CH2)CH2SiCl3
Formula Weight 322.98
Physical Form Liquid
Percent Purity ≥95.0%
Chemical Name or Material 1, 1-Bis(trichlorosilylmethyl)ethylene

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA