Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(1-Boc-4-piperidinyl)-4-iodopyrazole, tech. 90%

Click to view available options
Quantity:
250mg
1g
5g
877399-73-0
C13H20IN3O2
377.226
MFCD12913927
IONGFFYQZADMPH-UHFFFAOYSA-N
tert-butyl 4-4-iodo-1h-pyrazol-1-yl piperidine-1-carboxylate,1-boc-4-4-iodo-1h-pyrazol-1-yl piperidine,tert-butyl 4-4-iodopyrazol-1-yl piperidine-1-carboxylate,1-1-boc-4-piperidinyl-4-iodopyrazole,tert-butyl4-4-iodo-1h-pyrazol-1-yl piperidine-1-carboxylate,1-piperidinecarboxylic acid, 4-4-iodo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester,1-n-boc-4-4-iodopyrazol-1-yl piperidine,1-4-boc-piperidino-4-iodopyrazole,1-n-boc-4-4-iodo-1h-pyrazol-1-yl piperidine,1-1-tert-butoxycarbonyl-4-piperidinyl-4-iodopyrazole
45480389
tert-butyl 4-(4-iodopyrazol-1-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)N2C=C(C=N2)I
877399-73-0
C13H20IN3O2
377.226
MFCD12913927
IONGFFYQZADMPH-UHFFFAOYSA-N
tert-butyl 4-4-iodo-1h-pyrazol-1-yl piperidine-1-carboxylate,1-boc-4-4-iodo-1h-pyrazol-1-yl piperidine,tert-butyl 4-4-iodopyrazol-1-yl piperidine-1-carboxylate,1-1-boc-4-piperidinyl-4-iodopyrazole,tert-butyl4-4-iodo-1h-pyrazol-1-yl piperidine-1-carboxylate,1-piperidinecarboxylic acid, 4-4-iodo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester,1-n-boc-4-4-iodopyrazol-1-yl piperidine,1-4-boc-piperidino-4-iodopyrazole,1-n-boc-4-4-iodo-1h-pyrazol-1-yl piperidine,1-1-tert-butoxycarbonyl-4-piperidinyl-4-iodopyrazole
45480389
tert-butyl 4-(4-iodopyrazol-1-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)N2C=C(C=N2)I
Sensitivity Light Sensitive
Quantity 1g
Beilstein 14511019
Formula Weight 377.23
Physical Form Solid
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-(1-Boc-4-piperidinyl)-4-iodopyrazole