Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Cyclobutanedicarboxylatodiammineplatinum(II), ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
41575-94-4
C6H10N2O4Pt
369.24
MFCD00070464
YAYRGNWWLMLWJE-UHFFFAOYSA-L
carboplatin
122130863
azanide;cyclobutane-1,1-dicarboxylate;platinum(4+)
C1CC(C1)(C(=O)[O-])C(=O)[O-].[NH2-].[NH2-].[Pt+4]
41575-94-4
C6H10N2O4Pt
369.24
MFCD00070464
YAYRGNWWLMLWJE-UHFFFAOYSA-L
carboplatin
122130863
azanide;cyclobutane-1,1-dicarboxylate;platinum(4+)
C1CC(C1)(C(=O)[O-])C(=O)[O-].[NH2-].[NH2-].[Pt+4]
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Merck Index 15, 1823
Formula Weight 371.26
Physical Form Crystalline
Chemical Name or Material 1, 1-Cyclobutanedicarboxylatodiammineplatinum(II)

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause genetic defects.
Suspected of causing cancer.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage the

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.