Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane, 75% solution in aromatic free mineral spirit, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 349772500

Catalog No. AC349772500


Order in Bulk

Request bulk or custom formats

Add to Cart

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane
75% Sol.in Arom.free Min.spirit
74.0, 24.0
C2H4Cl2
MFCD00084801
ethylidene chloride, ethane, 1,1-dichloro, ethylidene dichloride, dichloroethane, 1,1-dichlorethane, aethylidenchlorid, 1,1-dicloroetano, 1,1-ethylidene dichloride, dichloromethylmethane, rcra waste number u076
CC(Cl)Cl
98.954
CHEBI:81599
74 to 76%
250 mL
Undesignated
75-34-3, 67-56-1
76.0, 26.0
[(CH3)3COO]2C6H7(CH3)3
0.8950g/mL
SCYULBFZEHDVBN-UHFFFAOYSA-N
1,1-dichloroethane
6365
302.45
Plastic bottle
Solution
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

GHS H Statement:
Heating may cause a fire.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

RUO – Research Use Only