Alfa Aesar™ 1,1-Dichloro-2-phenylcyclopropane, 97%

Manufacturer: Alfa Aesar™ A1227922

Quantity 100g

 View more versions of this product

Catalog No. AAA1227922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2415-80-7
    Molecular Formula C9H8Cl2
    Formula Weight 187.07
    MDL Number MFCD00001272
    Beilstein 1860351
    Synonym (2,2-Dichlorocyclopropyl)benzene
    Chemical Name or Material 1,1-Dichloro-2-phenylcyclopropane

    Specifications

    Formula Weight 187.07
    Molecular Formula C9H8Cl2
    CAS 2415-80-7
    Flash Point 104°C (219°F)
    Quantity 100g
    MDL Number MFCD00001272
    Synonym (2,2-Dichlorocyclopropyl)benzene
    Boiling Point 105° to 106°C (12mmHg)
    Beilstein 1860351
    Chemical Name or Material 1,1-Dichloro-2-phenylcyclopropane
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit