Alfa Aesar™ 1,1'-Diethyl-2,2'-cyanine iodide, 97%

Manufacturer: Alfa Aesar™ H5502203

Quantity 1g

 View more versions of this product

Catalog No. AAH5502203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 977-96-8
    Molecular Formula C23H23IN2
    Formula Weight 454.35
    MDL Number MFCD00011971
    UN Number UN2811
    Synonym Pseudoisocyanine iodide
    Chemical Name or Material 1,1'-Diethyl-2,2'-cyanine iodide

    Specifications

    Formula Weight 454.35
    TSCA TSCA
    Molecular Formula C23H23IN2
    MDL Number MFCD00011971
    CAS 977-96-8
    UN Number UN2811
    Melting Point 273°C
    Synonym Pseudoisocyanine iodide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H311-H330-H315-H319-H335
    Fatal if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 1,1'-Diethyl-2,2'-cyanine iodide
    Assay 97%

    Description

    Applications
    1,1 ft.-Diethyl-2,2 ft.-cyanine iodide is used to inhibit 1-methyl-4-phenylpyridinium (MPP+), toxic metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine that induces apoptosis in cerebellar granule neurons (CGNs).

    Solubility
    Soluble to 10 mM in dimethyl sulfoxide.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

    GHS H Statement
    H300-H311-H330-H315-H319-H335
    Fatal if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit