Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-diethylferrocene 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1273-97-8
C14H18Fe
MFCD01862455
1273-97-8
C14H18Fe
MFCD01862455
Color Brown-Red to Orange
Density 1.18g/mL
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5770 to 1.5800 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 242.14
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1, 1'-Diethylferrocene
Show More Show Less